N,N-Dicyclohexylmethylamine, 97%
N,N-Dicyclohexylmethylamine, 97%
N,N-Dicyclohexylmethylamine, 97%
Thermo Scientific Chemicals

N,N-Dicyclohexylmethylamine, 97%

CAS: 7560-83-0 | C13H25N | 195.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number 428390010
Price (USD)
208.00
Each
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
208.00
Each
Chemical Identifiers
CAS7560-83-0
IUPAC NameN-cyclohexyl-N-methylcyclohexanamine
Molecular FormulaC13H25N
InChI KeyGSCCALZHGUWNJW-UHFFFAOYSA-N
SMILESCN(C1CCCCC1)C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4890 to 1.4910 (20°C, 589 nm)
GC>=96.0 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only