4-Ethynylpyridine hydrochloride, 97%
4-Ethynylpyridine hydrochloride, 97%
4-Ethynylpyridine hydrochloride, 97%
Thermo Scientific Chemicals

4-Ethynylpyridine hydrochloride, 97%

CAS: 352530-29-1 | C7H5N·ClH | 139.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 428490050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS352530-29-1
IUPAC Name4-ethynylpyridine hydrochloride
Molecular FormulaC7H6ClN
InChI KeyKFBZWZGIZHLUBX-UHFFFAOYSA-N
SMILESCl.C#CC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
Appearance (Form)Crystalline powder
NMRconsistent with structure
Titration Argentometric>=96.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only