2-Naphthalenesulfonic acid hydrate, 90%, Technical
2-Naphthalenesulfonic acid hydrate, 90%, Technical
2-Naphthalenesulfonic acid hydrate, 90%, Technical
Thermo Scientific Chemicals

2-Naphthalenesulfonic acid hydrate, 90%, Technical

CAS: 76530-12-6 | C10H8O3S | 208.23 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4286900505 gGlass bottle
Catalog number 428690050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS76530-12-6
IUPAC Namenaphthalene-2-sulfonic acid
Molecular FormulaC10H8O3S
InChI KeyKVBGVZZKJNLNJU-UHFFFAOYSA-N
SMILESOS(=O)(=O)C1=CC=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey
HPLC>=88 %
Appearance (Form)Powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only