2-Methoxyethanol, for HPLC
2-Methoxyethanol, for HPLC
2-Methoxyethanol, for HPLC
Thermo Scientific Chemicals

2-Methoxyethanol, for HPLC

CAS: 109-86-4 | C3H8O2 | 76.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Catalog number 428780010
Price (USD)
149.00
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
149.00
Each
Chemical Identifiers
CAS109-86-4
IUPAC Name2-methoxyethan-1-ol
Molecular FormulaC3H8O2
InChI KeyXNWFRZJHXBZDAG-UHFFFAOYSA-N
SMILESCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 400 nm A: =<0.01
Water=<0.05 %
Residue after evaporation=<0.001 %
UV1 cm cell vs HPLC-grade water
UVat 220 nm A: =<1.0
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only