Potassium tert-butoxide, pure, 1.6-1.7M (20 wt.%) solution in THF, AcroSeal™
Potassium tert-butoxide, pure, 1.6-1.7M (20 wt.%) solution in THF, AcroSeal™
Potassium tert-butoxide, pure, 1.6-1.7M (20 wt.%) solution in THF, AcroSeal™
Potassium tert-butoxide, pure, 1.6-1.7M (20 wt.%) solution in THF, AcroSeal™
Thermo Scientific Chemicals

Potassium tert-butoxide, pure, 1.6-1.7M (20 wt.%) solution in THF, AcroSeal™

CAS: 865-47-4 | C4H9KO | 112.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 428791000
Price (USD)
156.00
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
156.00
Each
Chemical Identifiers
CAS865-47-4
IUPAC Namepotassium 2-methylpropan-2-olate
Molecular FormulaC4H9KO
InChI KeyLPNYRYFBWFDTMA-UHFFFAOYSA-N
SMILES[K+].CC(C)(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to amber
Strength1.60 to 1.75 M
AppearanceMay develop some turbidity or precipitate
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only