Benzyl chloroformate, 3M (50 wt.%) solution in toluene, AcroSeal™
Benzyl chloroformate, 3M (50 wt.%) solution in toluene, AcroSeal™
Benzyl chloroformate, 3M (50 wt.%) solution in toluene, AcroSeal™
Benzyl chloroformate, 3M (50 wt.%) solution in toluene, AcroSeal™
Thermo Scientific Chemicals

Benzyl chloroformate, 3M (50 wt.%) solution in toluene, AcroSeal™

CAS: 501-53-1 | C8H7ClO2 | 170.6 g/mol
Have Questions?
Catalog NumberQuantityPackaging
428901000100 mLAcroSeal™ Glass bottle
Catalog number 428901000
Price (USD)
66.65
Online Exclusive
73.80
Save 7.15 (10%)
Each
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)
66.65
Online Exclusive
73.80
Save 7.15 (10%)
Each
Add to cart
Chemical Identifiers
CAS501-53-1
IUPAC Namebenzyl carbonochloridate
Molecular FormulaC8H7ClO2
InChI KeyHSDAJNMJOMSNEV-UHFFFAOYSA-N
SMILESClC(=O)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear light yellow to light pink
Appearance (Form)Solution
GC45 to 53 % w/w
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only