Tetrabutylammonium hydroxide, 1M solution in methanol, AcroSeal™
Tetrabutylammonium hydroxide, 1M solution in methanol, AcroSeal™
Tetrabutylammonium hydroxide, 1M solution in methanol, AcroSeal™
Tetrabutylammonium hydroxide, 1M solution in methanol, AcroSeal™
Thermo Scientific Chemicals

Tetrabutylammonium hydroxide, 1M solution in methanol, AcroSeal™

CAS: 2052-49-5 | C16H37NO | 259.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 428951000
Price (USD)
145.65
Special offer
Online exclusive
Ends: 28-Dec-2026
171.00
Save 25.35
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
145.65
Special offer
Online exclusive
Ends: 28-Dec-2026
171.00
Save 25.35
Each
Chemical Identifiers
CAS2052-49-5
IUPAC Nametetrabutylazanium hydroxide
Molecular FormulaC16H37NO
InChI KeyVDZOOKBUILJEDG-UHFFFAOYSA-M
SMILES[OH-].CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HCl0.95 to 1.10 M
Appearance (Color)Clear colorless to yellow
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only