Titration Argentometric75 to 85 % (Product in toluene)
Appearance (Form)Solution
Refractive index1.4910 to 1.4970 (20°C, 589 nm)
Stabilizer0.3 % MgO
Appearance (Color)Colorless to yellow-orange
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.