Propylene carbonate, 99.5%, anhydrous, AcroSeal™
Propylene carbonate, 99.5%, anhydrous, AcroSeal™
Propylene carbonate, 99.5%, anhydrous, AcroSeal™
Propylene carbonate, 99.5%, anhydrous, AcroSeal™
Propylene carbonate, 99.5%, anhydrous, AcroSeal™
Thermo Scientific Chemicals

Propylene carbonate, 99.5%, anhydrous, AcroSeal™

CAS: 108-32-7 | C4H6O3 | 102.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 429261000
Price (USD)
58.65
Online exclusive
65.10
Save 6.45
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
58.65
Online exclusive
65.10
Save 6.45
Each
Chemical Identifiers
CAS108-32-7
IUPAC Name4-methyl-1,3-dioxolan-2-one
Molecular FormulaC4H6O3
InChI KeyRUOJZAUFBMNUDX-UHFFFAOYNA-N
SMILESCC1COC(=O)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=99.4 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
Water=<0.005 %

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only