Trimethyl orthoformate, 99+%, anhydrous, AcroSeal™
Trimethyl orthoformate, 99+%, anhydrous, AcroSeal™
Trimethyl orthoformate, 99+%, anhydrous, AcroSeal™
Trimethyl orthoformate, 99+%, anhydrous, AcroSeal™
Thermo Scientific Chemicals

Trimethyl orthoformate, 99+%, anhydrous, AcroSeal™

CAS: 149-73-5 | C4H10O3 | 106.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Catalog number 429281000
Price (USD)
37.65
Online exclusive
42.00
Save 4.35
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
37.65
Online exclusive
42.00
Save 4.35
Each
Chemical Identifiers
CAS149-73-5
IUPAC Nametrimethoxymethane
Molecular FormulaC4H10O3
InChI KeyPYOKUURKVVELLB-UHFFFAOYSA-N
SMILESCOC(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=99.0 %
Residue after evaporation=<0.005 %
Infrared spectrumConforms
Appearance (Color)Clear colorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only