2-chloropyrimidine-5-sulfonyl chloride, 97%
2-chloropyrimidine-5-sulfonyl chloride, 97%
2-chloropyrimidine-5-sulfonyl chloride, 97%
Thermo Scientific Chemicals

2-chloropyrimidine-5-sulfonyl chloride, 97%

CAS: 98026-88-1 | C4H2Cl2N2O2S | 213.03 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4296300101 gGlass bottle
Catalog number 429630010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS98026-88-1
IUPAC Name2-chloropyrimidine-5-sulfonyl chloride
Molecular FormulaC4H2Cl2N2O2S
InChI KeyDMAOAJYJIATWJS-UHFFFAOYSA-N
SMILESClC1=NC=C(C=N1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline solid
Melting point64°C to 68°C
Appearance (Color)White to yellow to light orange
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only