2-chloro-4-fluorophenylboronic acid, 97%
2-chloro-4-fluorophenylboronic acid, 97%
2-chloro-4-fluorophenylboronic acid, 97%
Thermo Scientific Chemicals

2-chloro-4-fluorophenylboronic acid, 97%

CAS: 313545-72-1 | C6H5BClFO2 | 174.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 430190010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS313545-72-1
IUPAC Name(2-chloro-4-fluorophenyl)boronic acid
Molecular FormulaC6H5BClFO2
InChI KeyXOFNMNLYGPKKOV-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(F)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only