Sequence Detection Primer, 80,000 pmol - FAQs
Sequence Detection Primer, 80,000 pmol - FAQs
View additional product information for Sequence Detection Primer - FAQs (4304970, 4304971, 4304972)
3 product FAQs found
I have purchased custom oligos (primers and probes) and I want to make a 20x working stock for a TaqMan assay, similar to the pre-designed assays. How do I dilute and mix my oligos to make a 20x working stock?
Most pre-designed TaqMan assays are supplied as a 20x assay concentrate containing 18 uM of each primer and 5 uM of probe which provides a 900 nM of each primer and 250 nM of the probe (for gene expression assays) and 200 nM of each probe (for SNP genotyping assays) at the final 1x concentration.
In order to make a 20x working stock from custom oligos, the oligos need to be diluted as follows: To make a 20x Assay stock for Gene Expression assays, make 100 uM stocks of both forward and reverse primers, and TaqMan probe. Add 18 ul of each primer and 5 ul of the TaqMan probe, which gives 41 ul volume. Add 59 ul of 1X TE to bring the volume to 100 ul of 20X TaqMan Gene Expression Assay. To make a 20x Assay stock for a Genotyping Assay, make 100 uM stocks of both forward and reverse primers, and both TaqMan probes. Add 18 ul of each primer and 4 ul of each TaqMan probe, which gives 44 ul volume. Add 56 ul of 1X TE to bring the volume to 100 ul of 20X TaqMan SNP Genotyping Assays.
What is the difference between Custom Standard DNA Oligos and Sequence Detection Primer (Cat. No. 4304970)?
The main difference between Custom Standard DNA Oligos and Sequence Detection Primer (Cat. Nos. 4304970, 4304971, 4304972) is that Sequence Detection Primers come in the guaranteed specific amount and concentration that is ordered. Whereas Custom DNA Oligos can be selected by the Synthesis Scale. The scale of synthesis (25 nmol, 50 nmol, 200 nmol, 1 µmol, or 10 µmol) refers to the starting scale and not the guaranteed final amount.
You can find more information about the Synthesis Scale by going to the Custom Standard DNA Oligos page and clicking on the question mark next to “Synthesis Scale.”
In addition, the sequence detection primers are desalted, whereas the custom DNA oligos can be purified by HPLC, PAGE, and Cartridge.
Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.