Total RNA Lysis Solution, Nucleic Acid Purification
Total RNA Lysis Solution is a 2X concentrate ideal for the chemical disruption of cells and tissue (cultured cells, primaryRead more
Have Questions?
Catalog Number
Quantity
4305895
250 mL
Catalog number 4305895
Price (USD)
209.00
Each
Quantity:
250 mL
Price (USD)
209.00
Each
Total RNA Lysis Solution is a 2X concentrate ideal for the chemical disruption of cells and tissue (cultured cells, primary cell isolates, and animal/plant tissues) prior to RNA or DNA extraction. Following downstream purification (e.g. using the 6100 PrepStation), intact RNA can be isolated that is free of contaminating protein and metal ions. Total RNA Lysis solution is the recommended reagent for the Total RNA Isolation Chemistry System for the ABI PRISM™ 6100 Nucleic Acid PrepStation.
For research use only. Not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration2X
For Use With (Equipment)6100 Nucleic Acid PrepStation