3-chloro-4-methylphenylboronic acid, 97%, Thermo Scientific Chemicals
3-chloro-4-methylphenylboronic acid, 97%, Thermo Scientific Chemicals
3-chloro-4-methylphenylboronic acid, 97%, Thermo Scientific Chemicals

3-chloro-4-methylphenylboronic acid, 97%, Thermo Scientific Chemicals

CAS: 175883-63-3 | Molecular Formula: C7H8BClO2 | Molecular Weight: 170.40 g/mol
Have Questions?
Catalog NumberQuantityPackaging
43062010010 gGlass bottle
Catalog number 430620100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS175883-63-3
IUPAC Name(3-chloro-4-methylphenyl)boronic acid
Molecular FormulaC7H8BClO2
InChI KeyYTJUYWRCAZWVSX-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1Cl)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder or flakes
Infrared spectrumConforms

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only