TaqMan™ Ribosomal RNA Control Reagents - FAQs
TaqMan™ Ribosomal RNA Control Reagents - FAQs
View additional product information for TaqMan™ Ribosomal RNA Control Reagents - FAQs (4308329)
2 product FAQs found
I am working on both mouse and human samples for gene expression studies. Do you have one endogenous control that I can use with both species?
The 18S rRNA is highly conserved among eukaryotes. We have several configurations of assays for this gene: P/N 4331182, Assay ID: Hs99999901_s1; PN 4319413E, 4310893E, 4333760T, 4333760F, 4308329. You can use any of these for both human and mouse samples, as well as samples from any other eukaryotic species.
What targets will the primer/probe set included in the TaqMan Ribosomal RNA Control Reagents kit (P/N 4308329) amplify?
The TaqMan Ribosomal RNA Control Reagents kit (P/N 4308329) will amplify all eukaryotic 18S rRNA sequences. For more information on this kit, please refer to the TaqMan Ribosomal RNA Control Reagents Protocol.