4309199 - FAQs
4309199 - FAQs
View additional product information for 4309199 - FAQs (4309199)
1 product FAQs found
Can Oligo d(T)16 RT primers be used with the TaqMan Human Endogenous Control Plate (P/N 4309199)?
No. The reverse transcription of total RNA to cDNA must be done with random hexamers or random primers (or a mixture of random and oligo dT primers) because the 18S rRNA assay cannot evaluate poly A+ purified RNA samples as most of the ribosomal RNA has been removed. The plate has been designed to evaluate only total RNA.