Borane-N,N-diethylaniline complex, 97%, AcroSeal™
Borane-N,N-diethylaniline complex, 97%, AcroSeal™
Borane-N,N-diethylaniline complex, 97%, AcroSeal™
Borane-N,N-diethylaniline complex, 97%, AcroSeal™
Thermo Scientific Chemicals

Borane-N,N-diethylaniline complex, 97%, AcroSeal™

CAS: 13289-97-9 | C10H15BN | 160.05 g/mol
Have Questions?
Catalog NumberQuantityPackaging
430931000100 mLAcroSeal™ Glass Bottle
Catalog number 430931000
Price (USD)
-
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS13289-97-9
IUPAC NameN,N-diethylaniline boron
Molecular FormulaC10H15BN
InChI KeyKHYAFFAGZNCWPT-UHFFFAOYSA-N
SMILES[B].CCN(CC)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Titration>=96 % (Active hydrogen)
Infrared spectrumConforms
Refractive index1.5220 to 1.5250 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only