3-Iodopyridine-4-carboxylic acid, 97%
3-Iodopyridine-4-carboxylic acid, 97%
3-Iodopyridine-4-carboxylic acid, 97%
Thermo Scientific Chemicals

3-Iodopyridine-4-carboxylic acid, 97%

CAS: 57842-10-1 | C6H4INO2 | 249.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
View all
Glass bottle
Catalog number 430970010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS57842-10-1
IUPAC Name3-iodopyridine-4-carboxylic acid
Molecular FormulaC6H4INO2
InChI KeyMJSNRTLYWDXHAQ-UHFFFAOYSA-N
SMILESOC(=O)C1=C(I)C=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to beige
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %
Melting point240°C to 244°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only