Single-Channel 60-well Temperature Verification Kit for PCR 9700 System, 0.5 mL - FAQs
Single-Channel 60-well Temperature Verification Kit for PCR 9700 System, 0.5 mL - FAQs
View additional product information for Single-Channel 60-well Temperature Verification Kit for PCR 9700 System, 0.5 mL - FAQs (4309924)
1 product FAQs found
Do you offer a temperature verification service to verify my PCR/qPCR instrument performance?
Yes, we offer an on-site temperature verification service with additional features such as ramp rate testing. Please find more information regarding the service below:
https://www.thermofisher.com/us/en/home/products-and-services/services/instrument-qualification-services/compliance-and-validation/on-site-temperature-verification.html