5-Bromo-1-methyl-1H-indole-3-carboxylic acid, 97%
5-Bromo-1-methyl-1H-indole-3-carboxylic acid, 97%
5-Bromo-1-methyl-1H-indole-3-carboxylic acid, 97%
Thermo Scientific Chemicals

5-Bromo-1-methyl-1H-indole-3-carboxylic acid, 97%

CAS: 400071-95-6 | C10H8BrNO2 | 254.08 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4311700101 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 431170010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS400071-95-6
IUPAC Name5-bromo-1-methyl-1H-indole-3-carboxylic acid
Molecular FormulaC10H8BrNO2
InChI KeyWQZLFFJMRKTCMU-UHFFFAOYSA-N
SMILESCN1C=C(C(O)=O)C2=CC(Br)=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
HPLC>=96.0 %
Infrared spectrumConforms
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only