MicroAmp™ Optical Adhesive Film
Applied Biosystems™

MicroAmp™ Optical Adhesive Film

Applied Biosystems™ MicroAmp™ Optical Adhesive Film은 microplate에 적용 시 well 간 오염과 샘플 증발 확률을 줄입니다.• 압력 감지 접착제로자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호케이스당 수
4311971100 Covers
436095425 Covers
카탈로그 번호 4311971
제품 가격(KRW)
397,000
온라인 행사
Ends: 30-Sep-2026
466,000
할인액 69,000 (15%)
Each
케이스당 수:
100 Covers
제품 가격(KRW)
397,000
온라인 행사
Ends: 30-Sep-2026
466,000
할인액 69,000 (15%)
Each
Applied Biosystems™ MicroAmp™ Optical Adhesive Film은 microplate에 적용 시 well 간 오염과 샘플 증발 확률을 줄입니다.

• 압력 감지 접착제로 각 well의 밀봉 보장.
• 확실한 샘플 결과 확인이 가능함
• Press-N-Seal: plate에 맞게 놓은 후 누르면 밀봉됨
• 적용이 용이함-장갑에 붙지 않습니다.

귀중한 샘플 손실을 방지함
투명한 이 접착 필름으로 microplate well에 샘플을 밀봉하세요. 샘플 간 교차 오염 확률을 줄이고 Real-Time PCR 데이터의 일관성을 보장합니다.

사용이 용이함
간단히 접착 필름 뒷면을 벗기고 microplate 위에 단단히 붙이세요. 평소와 같이 microplate를 놓고 샘플을 확인할 수 있으며 샘플 오염 확률을 줄입니다. MicroAmp™ 96-well과 384-well plate에 맞게 제작함.
사양
용도(장비)2720 Thermal Cycler, 6100 Thermal Cycler, 7000 System, 7300 System, 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, GeneAmp 9700, QuantStudio™, SimpliAmp™ Thermal Cycler, StepOnePlus™, Veriti Dx Fast Thermal Cycler, Veriti Dx Thermal Cycler, Veriti Fast Thermal Cycler, Veriti Thermal Cycler, ViiA™ 7 Dx System, ViiA™ 7 System, ViiA™ 7 Dx Fast System, QuantStudio™ Dx, 7500 Fast Dx System, ViiA™ 7 Fast System, ProFlex™ PCR System
케이스당 수100 Covers
제품 유형Film, Seal, or Applicator
수량100 covers
배송 조건Room Temperature
접착제Pressure Sensitive
색상Optical
물질Polyester
웰 수96
Plate Compatibility96/384-well Plate
제품라인MicroAmp
Unit SizeEach
구성 및 보관
Includes: 100 Optical Adhesive Covers
Storage: Room temperature

자주 묻는 질문(FAQ)

How do I correctly apply MicroAmp Optical Adhesive Covers to an optical 96- or 384-well plate?

Please follow the instructions below for the proper application of MicroAmp Optical Adhesive Covers (Cat. Nos. 4311971, 4313663 and 4360954).

The MicroAmp Optical Adhesive Cover uses a pressure-sensitive adhesive backing to adhere the cover to the optical 96- or 384-well plate. It is imperative that you use enough force to activate the pressure-sensitive adhesive to prevent evaporation from the wells.
Please note: Users should not expect the Optical Adhesive Cover to be very sticky to the touch on the side that adheres to the optical plate.

The correct way to apply the cover is as follows:

1. Apply firm pressure when using the Adhesive Seal Applicator (Cat. No. 4333183) to press the cover onto the plate.

2. Run the applicator firmly from left to right several times. Run the applicator up and down several times.

3. Run the edge of the applicator around the border (following the red line outside the wells) of the cover.

4. Repeat the left to right and up and down motions to assure a good final seal.

5. Inspect each well to make sure that each well edges are imprinted on the optical adhesive cover.
Please Note: Using the Applicator's edge to seal between the wells is NOT recommended. It may actually pull the seal off the wells.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Which of your plastic consumables are compatible with the Applied Biosystems 7900HT Fast Real-Time PCR System?

There are many of our reagents and plastic consumables that can be used on the Applied Biosystems 7900HT Fast Real-Time PCR System. Here are a few examples to help you get started:
Cat. No. 4313663, MicroAmp Optical Adhesive Film Kit
Cat. No. 4311971, 4360954, MicroAmp Optical Adhesive Film
Cat. No. 4323032, MicroAmp Optical 8-Cap Strip
Cat. No. 4309849; 4326270, MicroAmp Optical 384-Well Reaction Plate with Barcode
Cat. No. 4306737; 4326659; 4314320, MicroAmp Optical 96-Well Reaction Plate with Barcode
Cat. No. 4346906, MicroAmp Fast Optical 96-Well Reaction Plate with Barcode, 0.1 mL
Cat. No. 4312063, Splash Free 96-Well Base

For more comprehensive lists of compatible plastics, enter "Real-Time PCR Plastics" in the main Search field on our website and follow the links.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Can the MicroAmp Optical Adhesive Film be used with MicroAmp Optical Tubes?

No. The Optical Adhesive Film is recommended for use only with the MicroAmp Optical 96-well Reaction Plate (with or without barcode) and MicroAmp 384-well Optical Reaction Plate.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

What is the difference between the MicroAmp Optical Adhesive Covers and the MicroAmp Optical Caps? Can they be used interchangeably?

The MicroAmp Optical Adhesive Covers (P/N 4311971) have a higher optical efficiency than the MicroAmp Optical Caps, (8 caps/strip) (P/N 4323032). They can be used instead of the MicroAmp Optical Caps only when using the MicroAmp 96-Well Reaction Plate with Barcode (P/N 4306737, 4309849) or the MicroAmp Optical 96-Well Reaction Plate (P/N N801-0560). The Optical Adhesive Covers cannot be used with the MicroAmp Optical Tubes.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.