Can I use an MicroAmp Optical Cover Compression Pad on an Optical Fast 96-Well plate?
No. we do not recommend placing compression pads on the Fast 96-well plate or the StepOnePlus, ViiA 7, 7300, 7500, 7900HT, or 7500 Fast Real-Time PCR Systems.