MicroAmp™ Optical 96-Well Reaction Plate with Barcode & Optical Adhesive Films - FAQs
MicroAmp™ Optical 96-Well Reaction Plate with Barcode & Optical Adhesive Films - FAQs
View additional product information for MicroAmp™ Optical 96-Well Reaction Plate with Barcode & Optical Adhesive Films - FAQs (4314320)
3 product FAQs found
Can I place a standard MicroAmp 96-Well Optical Reaction Plate onto a 7500 Fast Real-Time PCR System?
No. The plate does not fit and will result in the crushing of the wells and possible contamination of the block due to leaking of the well contents.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Which of your plastic consumables are compatible with the Applied Biosystems 7900HT Fast Real-Time PCR System?
There are many of our reagents and plastic consumables that can be used on the Applied Biosystems 7900HT Fast Real-Time PCR System. Here are a few examples to help you get started:
Cat. No. 4313663, MicroAmp Optical Adhesive Film Kit
Cat. No. 4311971, 4360954, MicroAmp Optical Adhesive Film
Cat. No. 4323032, MicroAmp Optical 8-Cap Strip
Cat. No. 4309849; 4326270, MicroAmp Optical 384-Well Reaction Plate with Barcode
Cat. No. 4306737; 4326659; 4314320, MicroAmp Optical 96-Well Reaction Plate with Barcode
Cat. No. 4346906, MicroAmp Fast Optical 96-Well Reaction Plate with Barcode, 0.1 mL
Cat. No. 4312063, Splash Free 96-Well Base
For more comprehensive lists of compatible plastics, enter "Real-Time PCR Plastics" in the main Search field on our website and follow the links.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Can the MicroAmp Optical Adhesive Film be used with MicroAmp Optical Tubes?
No. The Optical Adhesive Film is recommended for use only with the MicroAmp Optical 96-well Reaction Plate (with or without barcode) and MicroAmp 384-well Optical Reaction Plate.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.