Product Image
Applied Biosystems™

Plate Septa, 384 well

These are septa for use with the 3730, 3730xl, 3130xl, 3130, 3100 and 3100-Avant™ instruments.
Have Questions?
Catalog NumberQuantity
431593420 each
Catalog number 4315934
Price (MXN)
-
Quantity:
20 each
These are septa for use with the 3730, 3730xl, 3130xl, 3130, 3100 and 3100-Avant™ instruments.

Use with:
384-Well Plate Base and 384-Well Plate Retainer

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)3100-Avant Genetic Analyzer
Format384-well Plate
Product TypeSepta Plate
Quantity20 each
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Each package contains 20 septa, sufficient for 7,680 samples.

Frequently asked questions (FAQs)

How often can the plate septa on the 3130/xl, 3730/xl, 3500/xl, and SeqStudio instruments be used?

We officially recommend that the plate septa be used one time only. However, we are aware that some of our customers will rinse the septa with water and re-use them. In those instances, the septa can become sticky and gummy over time and eventually start sticking to the array. In the best case scenario, the array will lift the plate off of the autosampler just enough to break the connection with the plate sensor and abort the run. The Event Log might show that the run aborted due to the user aborting it, so the run will appear to have completed or stopped with no errors and no data. The worst case scenario is that the plate remains on the array as the autosampler starts to move, bending the electrodes and breaking the array. We recommend replacing the plate septa regularly (replacing after each run is recommended). Do not use bleach, detergents, or solvents to clean the septa, it will cause them to dry out faster and also risks contaminating the system.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.