With the Oncomine Focus Assay, Chef-Ready Library kit, where can I find the combined barcoded AmpliSeq libraries when the run is complete?
After the completion of a run, the Ion Chef Instrument holds the barcoded libraries in the tube loaded in Position D of the Reagents cartridge. To avoid fluid loss due to evaporation, remove and cap the tube of combined barcoded libraries as soon as possible after run completion. Do not leave the tube in the instrument longer than 24 hours after the start of the run. After 24 hours from the start of the run, the instrument chiller will stop actively cooling, and the sample will be held at 27 degrees C.
The tube in Position D of the Ion AmpliSeq Chef Reagents DL8 Cartridge will contain 700 µL of combined barcoded libraries at completion of the run. The libraries are at >>100 pM (total combined library concentration) and are ready to use in template preparation. One can store unused portions of combined libraries at 4 to 8 degrees C for up to 1 month. For longer-term storage, store at -30 to -10 degrees C.
The tube in Position D of the Ion AmpliSeq Chef Reagents DL8 Cartridge will contain 700 µL of combined barcoded libraries at completion of the run. The libraries are at >>100 pM (total combined library concentration) and are ready to use in template preparation. One can store unused portions of combined libraries at 4 to 8 degrees C for up to 1 month. For longer-term storage, store at -30 to -10 degrees C.