TaqMan™ Universal PCR Master Mix, 10 x 5 mL - FAQs
TaqMan™ Universal PCR Master Mix, 10 x 5 mL - FAQs
View additional product information for TaqMan™ Universal PCR Master Mix - FAQs (4305719, 4364338, 4326708, 4318157, 4304437, 4364340)
1 product FAQs found
What master mix can I use with the TaqMan SNP Genotyping Assays?
We recommend using either the TaqPath ProAmp Master Mix or the TaqMan Genotyping Master Mix. The TaqMan Genotyping Master Mix has the advantage of proven performance with up to 3 days of pre- and post-PCR stability, allowing you to set up plates ahead of time or read the plates later (see the data here, https://tools.thermofisher.com/content/sfs/brochures/cms_039236.pdf) while the TaqPath ProAmp Master Mix can handle samples that may have inhibitors present.