N,N-Dimethylacrylamide, 99%, stabilized with 500 ppm MEHQ
N,N-Dimethylacrylamide, 99%, stabilized with 500 ppm MEHQ
N,N-Dimethylacrylamide, 99%, stabilized with 500 ppm MEHQ
Thermo Scientific Chemicals

N,N-Dimethylacrylamide, 99%, stabilized with 500 ppm MEHQ

CAS: 2680-03-7 | C5H9NO | 99.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 432021000
Price (USD)
72.60
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
72.60
Each
Chemical Identifiers
CAS2680-03-7
IUPAC NameN,N-dimethylprop-2-enamide
Molecular FormulaC5H9NO
InChI KeyYLGYACDQVQQZSW-UHFFFAOYSA-N
SMILESCN(C)C(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Refractive index1.4730 to 1.4750 (20°C, 589 nm)
Infrared spectrumConforms
StabilizerApprox. 500 ppm
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only