Tetratriacontane, 98%
Tetratriacontane, 98%
Tetratriacontane, 98%
Thermo Scientific Chemicals

Tetratriacontane, 98%

CAS: 14167-59-0 | C34H70 | 478.93 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4320700101 gGlass bottle
Catalog number 432070010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS14167-59-0
IUPAC Nametetratriacontane
Molecular FormulaC34H70
InChI KeyGWVDBZWVFGFBCN-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point72°C to 76°C
Elemental Analysis14.36 to 15.10 % H
Appearance (Form)Flakes
Appearance (Color)White
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only