How many SNPs can the SNaPshot Multiplex kit analyze?
The SNaPshot Multiplex Kis is designed to interrogate up to 10 single nucleotide polymorphisms (SNPs) at known locations on 1-10 templates in a single tube.
Search
View additional product information for SNaPshot™ Multiplex Kit - FAQs (4323159, 4323163, 4323151, 4323161)
4 product FAQs found