Methyl 3-aminobenzo[b]thiophene-2-carboxylate, 97%
Methyl 3-aminobenzo[b]thiophene-2-carboxylate, 97%
Methyl 3-aminobenzo[b]thiophene-2-carboxylate, 97%
Thermo Scientific Chemicals

Methyl 3-aminobenzo[b]thiophene-2-carboxylate, 97%

CAS: 35212-85-2 | C10H9NO2S | 207.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 432410010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS35212-85-2
IUPAC Namemethyl 3-amino-1-benzothiophene-2-carboxylate
Molecular FormulaC10H9NO2S
InChI KeyVLHHEYMZLXKSQO-UHFFFAOYSA-N
SMILESCOC(=O)C1=C(N)C2=CC=CC=C2S1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)Yellow to brown
Infrared spectrumConforms
HPLC>=96.0 %
Melting point108°C to 112°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only