N-Methylmorpholine, 99.5%, purified by redistillation, AcroSeal™
N-Methylmorpholine, 99.5%, purified by redistillation, AcroSeal™
N-Methylmorpholine, 99.5%, purified by redistillation, AcroSeal™
Thermo Scientific Chemicals

N-Methylmorpholine, 99.5%, purified by redistillation, AcroSeal™

CAS: 109-02-4 | C5H11NO | 101.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 432800010
Price (EUR)
82,10
Each
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (EUR)
82,10
Each
Chemical Identifiers
CAS109-02-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC>=99.4 %
Infrared spectrumConforms
Water=<0.5 %
Color scale=<20 APHA
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only