Sodium (para)periodate, 98%
Sodium (para)periodate, 98%
Sodium (para)periodate, 98%
Thermo Scientific Chemicals

Sodium (para)periodate, 98%

CAS: 13940-38-0 | H2INa3O6 | 293.88 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 432855000
Price (EUR)
300,00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
300,00
Each
Specifications
Assay Percent Range97.5% min. (Iodometry)
CAS13940-38-0
Chemical Name or MaterialSodium (para)periodate
ColorWhite
EINECS Number237-720-1
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only