This product was discontinued in 2012. We recommend QuantStudio qPCR product family. Learn more
7900HT System Automation Accessory Upgrade
Applied Biosystems™
7900HT System Automation Accessory Upgrade
The 7900HT System Automation Accessory Upgrade is an optional accessory for the 7900HT Fast Real-Time PCR System.The 7900HT Fast Real-TimeRead more
Have Questions?
Catalog NumberQuantity
43290071 upgrade
Catalog number 4329007
Price (USD)
-
Quantity:
1 upgrade
The 7900HT System Automation Accessory Upgrade is an optional accessory for the 7900HT Fast Real-Time PCR System.
The 7900HT Fast Real-Time PCR System is the only real-time quantitative PCR system that combines 96- and 384-well plate compatibility and the TaqMan™ Low Density Array with fully automated robotic loading-and now also offers optional Fast real-time PCR capability. The optional, integrated Automation Accessory provides maximum throughput. Up to eighty-four 384-well plates can be loaded at a single time for unattended operation; a bar code reader allows you to insert or substitute plates into the sample queue at any time.
• Fast PCR option reduces run time to about 35 minutes in a standard 96-well format, or about 55 minutes in a 384-well plate.
• Continuous wavelength detection from 500-660 nm allows the use of multiple fluorophores in a single reaction.
• Measure gene expression levels, detect and quantitate pathogens, perform allelic discrimination (SNP genotyping) assays as well as score the presence of gene sequences.
• 96- or 384-well plate compatibility (including the TaqMan™ Low Density Array).
• Optional Enterprise edition software provides data analysis tools that support 21 CFR Part 11 guidelines providing data integrity and security.
• Hands-free plate-loading and unloading provides true walkaway automation allowing you to increase your lab's productivity.
• Proven assay development guidelines save time and money
High Throughput
The 7900HT System is a high-throughput real-time PCR system that detects and quantitates nucleic acid sequences. An Automation Accessory combined with 384-well plate capability make the 7900HT system ideally suited to meet the high-throughput requirements of today's drug discovery process. Key applications include gene expression quantitation and the detection of single nucleotide polymorphisms (SNPs) using the fluorogenic 5' nuclease assay.
Flexibility
The 7900HT system is a versatile research tool that can accommodate any real-time PCR need. User-interchangeable thermal cycling block formats let you select the format that’s right for your project, using industry-standard 96- and 384-well formats, as well as a novel 384-well TaqMan™ Low Density Array and a new Fast 96-well block that reduces run times from 2 hours to about 30 minutes. An easy automation upgrade path lets you add features to meet throughput demands. A Fast PCR option reduces run times from 2 hours to about 30 minutes.
Powerful Software
Using two automation software tools, Plate Utility and Automation Controller, most of the gene expression and SNP genotyping workflow is fully automated for high-throughput applications and requires minimal user intervention. You can now process 5,000 x 384 well plates (384 markers), which equates to 1.92 million genotypes. Using Relative Quantification (RQ) Manager, you can also process 200 x 384-well plates (96 detectors with quadruplicate data points⁄multiplexed endogenous control), which equates to 153,600 data points. Fully integrated into one complete Enterprise system, SNP Manager and RQ Manager eliminate data analysis bottlenecks in high-throughput SNP and gene expression research by allowing significantly more plates to be analyzed simultaneously. This reduces hands-on analysis time and simplifies the data analysis workflow.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Real Time PCR (qPCR)
Quantity1 upgrade
CapacityUp to 84 x 384-well Plates, Up to 36 x 96-well Plates
Product TypeSystem Upgrade
Unit SizeEach
Contents & Storage
1 Upgrade
Citations & References (4396)
Citations & References
Abstract
Increased hepatobiliary clearance of unconjugated thyroxine determines DMP 904-induced alterations in thyroid hormone homeostasis in rats.
Authors:Wong H; Lehman-McKeeman LD; Grubb MF; Grossman SJ; Bhaskaran VM; Solon EG; Shen HS; Gerson RJ; Car BD; Zhao B; Gemzik B
Journal:Toxicological Sciences : An Official Journal of the Society of Toxicology
PubMed ID:
4-(3-pentylamino)-2,7-dimethyl-8-(2-methyl-4-methoxyphenyl)-pyrazolo-[1,5-a]-pyrimidine (DMP 904) is a potent and selective antagonist of corticotropin releasing factor receptor-1 (CRF1 receptor) with an efficacious anxiolytic profile in preclinical animal models. In subchronic toxicity studies in Sprague-Dawley rats, DMP 904 produced thyroid follicular cell hypertrophy and hyperplasia, and a low incidence of follicular cell adenoma. The ... More
RAGE ligand upregulation of VEGF secretion in ARPE-19 cells.
Authors:Ma W; Lee SE; Guo J; Qu W; Hudson BI; Schmidt AM; Barile GR
Journal:Investigative Ophthalmology & Visual Science
PubMed ID:
PURPOSE: The importance of VEGF in stimulating neovascular age-related macular degeneration (AMD) is well-recognized, but the initiating factors that induce local upregulation of VEGF remain unclear. The current study was conducted to test the hypothesis that activation of RAGE (receptor for advanced glycation end products [AGEs]) by its ligands, including ... More
SEROTONIN POLYMORPHISMS AND POSTTRAUMATIC STRESS DISORDER IN A TRAUMA EXPOSED AFRICAN AMERICAN POPULATION
Authors:Mellman, TA; Alim, T; Brown, DD; Gorodetsky, E; Buzas, B; Lawson, WB; Goldman, D; Charney, DS
Journal:DEPRESSION AND ANXIETY
PubMed ID:
Background: Genetic polymorphisms that influence serotonin (5-hydroxytryptamine, 5HT) neurotransmission are candidates for contributing to susceptibility to posttraumatic stress disorder (PTSD). The objective of our study was to determine if a variable length polymorphism for the promoter regions of the 5HT transporter (5HTTLPR), and/or a substitution polymorphism. in the promoter region ... More
Cognitive Flexibility is Associated with KIBRA Variant and Modulated by Recent Tobacco Use
Authors:Zhang, HP; Kranzler, HR; Poling, J; Gruen, JR; Gelernter, J
Journal:NEUROPSYCHOPHARMACOLOGY
PubMed ID:
The kidney and brain expressed protein gene (KIBRA) and the calsyntenin 2 gene (CLSTN2) are reportedly involved in synaptic plasticity. Single nucleotide polymorphisms (SNPs) rs17070145 (KIBRA) and rs6439886 (CLSTN2) have been found to affect memory performance measures. This study examined the association of KIBRA SNP rs17070145 and CLSTN2 SNPs rs6439886 ... More
Phenomic determinants of genomic variation in autism spectrum disorders
Authors:Qiao, Y; Riendeau, N; Koochek, M; Liu, X; Harvard, C; Hildebrand, MJ; Holden, JJA; Rajcan-Separovic, E; Lewis, MES
Journal:Journal of Medical Genetics
PubMed ID:
Background: Autism spectrum disorders (ASDs) are common, heritable neurobiologic conditions of unknown aetiology confounded by significant clinical and genetic heterogeneity. Methods: This study evaluated a broad categorisation of phenotypic traits (or phenome) for 100 subjects with Autism Diagnostic Interview-Revised/Autism Diagnostic Observation Schedule-Generic (ADI-R/ADOS-G) confirmed idiopathic ASD undergoing 1 Mb ... More