I am observing unexpected peaks in my SNaPshot data. Why?
The extra peaks could be due to incomplete removal of PCR primers, incomplete removal of dNTPs from the PCR reaction, or incomplete removal of the fluorescently labeled ddNTPS from the SNaPshot reaction. Residual PCR primers and dNTPs will both participate in the SNaPshot reaction. For the PCR purification, please use fresh SAP and Exo1 or use another method of PCR purification. To address incomplete removal of fluorescently labeled ddNTPS, use fresh CIP or SAP.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.