2-Azetidinone, 98%
2-Azetidinone, 98%
2-Azetidinone, 98%
Thermo Scientific Chemicals

2-Azetidinone, 98%

CAS: 930-21-2 | C3H5NO | 71.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 432960010
Price (USD)
131.65
Online exclusive
146.00
Save 14.35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
131.65
Online exclusive
146.00
Save 14.35
Each
Chemical Identifiers
CAS930-21-2
IUPAC Nameazetidin-2-one
Molecular FormulaC3H5NO
InChI KeyMNFORVFSTILPAW-UHFFFAOYSA-N
SMILESO=C1CCN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Crystalline solid
Melting point72°C to 77°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only