Product Image
Applied Biosystems™

Tool, 5/16 HEX 6 in. L W/T-handle

This is a tool for use with the 7900HT Sequence Detection System.
Have Questions?
Catalog NumberQuantity
43302511 unit
Catalog number 4330251
Price (USD)
104.00
Each
Quantity:
1 unit
Price (USD)
104.00
Each

This is a tool for use with the 7900HT Sequence Detection System.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Application)Real Time PCR (qPCR)
For Use With (Equipment)Sequence Detection System
Quantity1 unit
Shipping ConditionRoom Temperature
Product TypeTool
Unit SizeEach
Contents & Storage
Store at Room Temperature