This product was discontinued in 2012. We recommend QuantStudio qPCR product family. Learn more
7900HT System Standard 96-Well Block Upgrade Kit
Applied Biosystems™

7900HT System Standard 96-Well Block Upgrade Kit

The 7900HT System Standard 96-Well Block Upgrade Kit is an optional accessory for the 7900HT Fast Real-Time PCR System, whichRead more
Have Questions?
Catalog NumberQuantity
43314051 kit
Catalog number 4331405
Price (USD)
-
Quantity:
1 kit
The 7900HT System Standard 96-Well Block Upgrade Kit is an optional accessory for the 7900HT Fast Real-Time PCR System, which is available is either a 96- or 384-well block format. The interchangeable 96- and 384-well block capability combined with the optional Automation Accessory offer unmatched flexibility. These features make the 7900HT system ideally suited to meet the high-throughput requirements of today’s drug discovery process. Key applications include gene expression quantitation and the detection of single nucleotide polymorphisms (SNPs) using the fluorogenic 5’ nuclease assay.

The 7900HT Fast Real-Time PCR System is the only real-time quantitative PCR system that combines 96- and 384-well plate compatibility and the TaqMan™ Low Density Array with fully automated robotic loading-and now also offers optional Fast real-time PCR capability.
• Fast PCR option reduces run time to about 35 minutes in a standard 96-well format, or about 55 minutes in a 384-well plate.
• Continuous wavelength detection from 500-660 nm allows the use of multiple fluorophores in a single reaction.
• Measure gene expression levels, detect and quantitate pathogens, perform allelic discrimination (SNP genotyping) assays as well as score the presence of gene sequences.
• 96- or 384-well plate compatibility (including the TaqMan™ Low Density Array).
• Optional Enterprise edition software provides data analysis tools that support 21 CFR Part 11 guidelines providing data integrity and security.
• Hands-free plate-loading and unloading provides true walkaway automation allowing you to increase your lab's productivity.
• Proven assay development guidelines save time and money

High Throughput
The 7900HT System is a high-throughput real-time PCR system that detects and quantitates nucleic acid sequences. An Automation Accessory combined with 384-well plate capability make the 7900HT system ideally suited to meet the high-throughput requirements of today's drug discovery process. Key applications include gene expression quantitation and the detection of single nucleotide polymorphisms (SNPs) using the fluorogenic 5' nuclease assay.

Flexibility
The 7900HT system is a versatile research tool that can accommodate any real-time PCR need. User-interchangeable thermal cycling block formats let you select the format that’s right for your project, using industry-standard 96- and 384-well formats, as well as a novel 384-well TaqMan™ Low Density Array and a new Fast 96-well block that reduces run times from 2 hours to about 30 minutes. An easy automation upgrade path lets you add features to meet throughput demands. A Fast PCR option reduces run times from 2 hours to about 30 minutes.

Powerful Software
Using two automation software tools, Plate Utility and Automation Controller, most of the gene expression and SNP genotyping workflow is fully automated for high-throughput applications and requires minimal user intervention. You can now process 5,000 x 384 well plates (384 markers), which equates to 1.92 million genotypes. Using Relative Quantification (RQ) Manager, you can also process 200 x 384-well plates (96 detectors with quadruplicate data points⁄multiplexed endogenous control), which equates to 153,600 data points. Fully integrated into one complete Enterprise system, SNP Manager and RQ Manager eliminate data analysis bottlenecks in high-throughput SNP and gene expression research by allowing significantly more plates to be analyzed simultaneously. This reduces hands-on analysis time and simplifies the data analysis workflow.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Real Time PCR (qPCR)
For Use With (Equipment)7900HT
Format96-well Plate
Quantity1 kit
Reaction SpeedStandard
Block FormatInterchangeable
Product TypeBlock Upgrade Kit
Unit SizeEach
Contents & Storage
1 Upgrade

Frequently asked questions (FAQs)

Can I import a standard curve for my real-time PCR data into the Design & Analysis Software (v2.x)?

It is possible to import a standard curve into the Design & Analysis Software (v2.x) if you are analyzing results from one of the following Applied Biosystems real-time PCR instruments:
• 7900HT Fast Real-Time PCR System
• 7500/7500 Fast Real-Time PCR System
• StepOne/StepOne Plus Real-Time PCR System
• ViiA 7 Real Time PCR System
• QuantStudio 1 Real-Time PCR System
• QuantStudio 3 & 5 Real-Time PCR System
• QuantStudio 6 & 7 Flex Real-Time PCR System
• QuantStudio 6 & 7 Pro Real-Time PCR Systems
• QuantStudio 12K Flex Real-Time PCR system

Follow the instructions below to import a standard curve for analysis in the Design & Analysis Software (v2.x):
1. Open the data file in the Design & Analysis Software (v2.x).
2. Click on Actions.
3. Click on "Standard Curve Analysis Setting".
4. Select "External Standard Curves" and click on "Import".
5. Browse to the proper .csv file and click "Open".
6. Click "Apply".

What is the difference between a Fast 96-well and standard 96-well block?

There are two main differences between a Fast 96-well and standard 96-well block for the Applied Biosystems real-time PCR systems:
• Reaction well volume: The Fast 96-well blocks have 0.1 mL reaction wells and are compatible with 0.1 mL plates and tubes. The standard 96-well blocks have 0.2 mL reaction wells and are compatible with 0.2 mL plates and tubes.
• Ramp rate: The fast 96-well blocks have a higher maximum ramp rate than standard blocks for the same real-time PCR system. However, both the fast 96-well and standard 96-well blocks are capable of running standard and fast chemistry.

Note: For newer instruments (QuantStudio 3 and 5 Real-Time PCR Systems and later) "0.1 mL" and "0.2 mL" are being used in place of "fast" and "standard" as designations for the two 96-well block formats.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Citations & References (4396)

Citations & References
Abstract
Increased hepatobiliary clearance of unconjugated thyroxine determines DMP 904-induced alterations in thyroid hormone homeostasis in rats.
Authors:Wong H; Lehman-McKeeman LD; Grubb MF; Grossman SJ; Bhaskaran VM; Solon EG; Shen HS; Gerson RJ; Car BD; Zhao B; Gemzik B
Journal:Toxicological Sciences : An Official Journal of the Society of Toxicology
PubMed ID:
4-(3-pentylamino)-2,7-dimethyl-8-(2-methyl-4-methoxyphenyl)-pyrazolo-[1,5-a]-pyrimidine (DMP 904) is a potent and selective antagonist of corticotropin releasing factor receptor-1 (CRF1 receptor) with an efficacious anxiolytic profile in preclinical animal models. In subchronic toxicity studies in Sprague-Dawley rats, DMP 904 produced thyroid follicular cell hypertrophy and hyperplasia, and a low incidence of follicular cell adenoma. The ... More
RAGE ligand upregulation of VEGF secretion in ARPE-19 cells.
Authors:Ma W; Lee SE; Guo J; Qu W; Hudson BI; Schmidt AM; Barile GR
Journal:Investigative Ophthalmology & Visual Science
PubMed ID:
PURPOSE: The importance of VEGF in stimulating neovascular age-related macular degeneration (AMD) is well-recognized, but the initiating factors that induce local upregulation of VEGF remain unclear. The current study was conducted to test the hypothesis that activation of RAGE (receptor for advanced glycation end products [AGEs]) by its ligands, including ... More
SEROTONIN POLYMORPHISMS AND POSTTRAUMATIC STRESS DISORDER IN A TRAUMA EXPOSED AFRICAN AMERICAN POPULATION
Authors:Mellman, TA; Alim, T; Brown, DD; Gorodetsky, E; Buzas, B; Lawson, WB; Goldman, D; Charney, DS
Journal:DEPRESSION AND ANXIETY
PubMed ID:
Background: Genetic polymorphisms that influence serotonin (5-hydroxytryptamine, 5HT) neurotransmission are candidates for contributing to susceptibility to posttraumatic stress disorder (PTSD). The objective of our study was to determine if a variable length polymorphism for the promoter regions of the 5HT transporter (5HTTLPR), and/or a substitution polymorphism. in the promoter region ... More
Cognitive Flexibility is Associated with KIBRA Variant and Modulated by Recent Tobacco Use
Authors:Zhang, HP; Kranzler, HR; Poling, J; Gruen, JR; Gelernter, J
Journal:NEUROPSYCHOPHARMACOLOGY
PubMed ID:
The kidney and brain expressed protein gene (KIBRA) and the calsyntenin 2 gene (CLSTN2) are reportedly involved in synaptic plasticity. Single nucleotide polymorphisms (SNPs) rs17070145 (KIBRA) and rs6439886 (CLSTN2) have been found to affect memory performance measures. This study examined the association of KIBRA SNP rs17070145 and CLSTN2 SNPs rs6439886 ... More
Phenomic determinants of genomic variation in autism spectrum disorders
Authors:Qiao, Y; Riendeau, N; Koochek, M; Liu, X; Harvard, C; Hildebrand, MJ; Holden, JJA; Rajcan-Separovic, E; Lewis, MES
Journal:Journal of Medical Genetics
PubMed ID:
Background: Autism spectrum disorders (ASDs) are common, heritable neurobiologic conditions of unknown aetiology confounded by significant clinical and genetic heterogeneity. Methods: This study evaluated a broad categorisation of phenotypic traits (or phenome) for 100 subjects with Autism Diagnostic Interview-Revised/Autism Diagnostic Observation Schedule-Generic (ADI-R/ADOS-G) confirmed idiopathic ASD undergoing 1 Mb ... More