POROS™ Heparin 50 μm Column, 4.6 x 100 mm, 1.7 mL - FAQs
POROS™ Heparin 50 μm Column, 4.6 x 100 mm, 1.7 mL - FAQs
View additional product information for POROS™ Heparin 50 μm Affinity Columns - FAQs (4333411, 4333413, 4333412)
This product does not have any FAQs available.