Product Image
Applied Biosystems™

96-Well and 384-Well Plate Retainer (heat seal)

This is a 96- and 384-well Plate Retainer (Heat Seal) for use with Applied Biosystems 3730 and 3730xl DNA Analyzers.
Have Questions?
Catalog NumberQuantity
43348654 each
Catalog number 4334865
Price (USD)
83.00
Each
Quantity:
4 each
Price (USD)
83.00
Each

This is a 96- and 384-well Plate Retainer (Heat Seal) for use with Applied Biosystems 3730 and 3730xl DNA Analyzers.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Compatibility96-Well Plates, 384-well plate, 96-well plate, 384-Well Plates
For Use With (Equipment)3730xl DNA Analyzer, 3730 DNA Analyzer
Format96-well Plate, 384-well Plate
Product TypePlate Retainer
Quantity4 each
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Each package contains 4 plate retainers.