What is the appropriate volume of liquid for use in the buffer, water, and waste reservoirs, on the Applied Biosystems 3730 Series DNA Analyzer?
The appropriate volume to use in the buffer, water, and waste reservoirs is 80mls. The black line on the reservoir body is a reference only and should not be used to determine the fill volume. For accuracy and reproducibility in filling the reservoirs, a graduated cylinder should be used each time the reservoir fluids are replaced.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.