2,4,6-Trifluorophenylboronic acid, 95%
2,4,6-Trifluorophenylboronic acid, 95%
2,4,6-Trifluorophenylboronic acid, 95%
Thermo Scientific Chemicals

2,4,6-Trifluorophenylboronic acid, 95%

CAS: 182482-25-3 | C6H4BF3O2 | 175.90 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 433590010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS182482-25-3
IUPAC Name(2,4,6-trifluorophenyl)boronic acid
Molecular FormulaC6H4BF3O2
InChI KeyIPEIGKHHSZFAEW-UHFFFAOYSA-N
SMILESOB(O)C1=C(F)C=C(F)C=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Appearance (Form)Crystalline powder
HPLC>=94.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only