2-Pyridylzinc bromide, 0.5M solution in THF, AcroSeal™
2-Pyridylzinc bromide, 0.5M solution in THF, AcroSeal™
2-Pyridylzinc bromide, 0.5M solution in THF, AcroSeal™
Thermo Scientific Chemicals

2-Pyridylzinc bromide, 0.5M solution in THF, AcroSeal™

CAS: 218777-23-2 | C5H4BrNZn | 223.39 g/mol
Have Questions?
Catalog NumberQuantityPackaging
43363050050 mLAcroSeal™ Glass Bottle
Catalog number 433630500
Price (USD)
188.65
Online exclusive
209.00
Save 20.35
Each
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
188.65
Online exclusive
209.00
Save 20.35
Each
Chemical Identifiers
CAS218777-23-2
IUPAC Namebromozincylium; pyridin-2-ide
Molecular FormulaC5H4BrNZn
InChI KeyZQJYTTPJYLKTTI-UHFFFAOYSA-M
SMILES[Zn+]Br.C1=CC=N[C-]=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark green to brown to black
Appearance (Form)Solution
Titration Complexometric0.45 to 0.65 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only