Product Image
Applied Biosystems™

BigDye™ Terminator v1.1 & v3.1 5X Sequencing Buffer

본 sequencing 버퍼는BigDye™ Terminator Cycle Sequencing Kit에 사용하도록 최적화되어 있습니다.이 제품은 연구용으로만 사용가능합니다. 진단 절차에 사용할 수 없습니다.자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
43366971 mL
433669928 mL
4336701233 mL
카탈로그 번호 4336697
제품 가격(KRW)
135,000
온라인 행사
Ends: 31-Dec-2026
150,000
할인액 15,000
Each
수량:
1 mL
제품 가격(KRW)
135,000
온라인 행사
Ends: 31-Dec-2026
150,000
할인액 15,000
Each
본 sequencing 버퍼는BigDye™ Terminator Cycle Sequencing Kit에 사용하도록 최적화되어 있습니다.

이 제품은 연구용으로만 사용가능합니다. 진단 절차에 사용할 수 없습니다.
For Research Use Only. Not for use in diagnostic procedures.
사양
버퍼Sequencing Buffer
농도5X
용도(애플리케이션)Standard Sequencing
용도(장비)GeneAmp 9700, Veriti Thermal Cycler, 3500 Genetic Analyzer, 3730xl DNA Analyzer, 3130xl Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer
형태Liquid
제품라인BigDye
제품 유형Sequencing Buffer
수량1 mL
배송 조건Room Temperature
Unit SizeEach
구성 및 보관
Store at 2°C to 8°C.

자주 묻는 질문(FAQ)

I am getting shorter reads than I used to. What can cause this?

A change in the read length is usually caused by something that either changes the migration of the DNA or the efficiency of the sequencing reaction. Some of the things that can cause this are:

• Buffer: Buffer may be prepared incorrectly or on the system for too long.

• Polymer: Polymer may have been on the system the system too long, used past the expiration date, frozen/crystallized and thawed, or mixed with water or buffer during capillary fill. There may be a mismatch between polymer type selected in software and what is installed on the system.

• Environment: Instrument may be operating outside environmental specifications in the Site Preparation Guide. You may have poor/improper instrument ventilation, or air flow may be blowing directly on the system.

• DNA quality: RNA/protein contamination may be reducing the sequencing reaction efficiency. If working with PCR product, there may be carryover of primers and/or dNTPs from the PCR reaction, or the sequencing reaction cleanup is incomplete. If using a kit that has beads in it for the purification, beads may be blocking the capillary tip. If using BigDye XTerminator Purification, heating the XTerminator reagent can cause loss of smaller products.

• DNA quantity: There may be too much DNA competing for entry into the capillary with labeled product, or excessive amounts of DNA creating a temporary blockage of the capillary.

For more information on other causes of short reads and how to address each issue, please refer to the DNA Sequencing by Capillary Electrophoresis: Applied Biosystems Chemistry Guide: Second Edition (Cat. No. 4305080, Rev. C). The guide can be found by searching the Thermo Fisher Scientific website with the catalog number 4305080.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

Can I dilute my BigDye Terminator reactions?

The BigDye Terminator v1.1 and v3.1 chemistries can be diluted, and a 5X Sequencing Buffer is provided with the kit to enable you to do so. However, dilution of the BigDye Terminator may compromise your data integrity depending on the cycle sequencing protocol you are following and the sequence of the particular DNA you are investigating. TheBigDye Direct Cycle Sequencing Kit and the dGTP BigDye Terminator Ready Reaction Kits v1.0 and v3.0 cannot be diluted.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

I received BigDye Terminator v1.1 & v3.1 5X Sequencing Buffer (Cat. No. 4336697) at room temperature. Is this correct?

Yes, BigDye Terminator v1.1 & v3.1 5X Sequencing Buffer is shipped at room temperature and needs to be stored at 2-8 degrees C upon receipt.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

How do I calculate the amount of BigDye Terminator v1.1 & v3.1 5X Buffer to use in the reaction?

The calculation can be done manually or by using our Applied Biosystems BigDye Terminator v1.1 & v3.1 5X Sequencing Buffer Calculator.

The formula is provided below:

Volume of BigDye Terminator v1.1 & v3.1 5X Buffer (µL) = 0.5 x [(total reaction volume/2.5) - volume of BigDye Terminator Ready Reaction Mix].

For example, if you are using 2 µL of the BigDye Terminator Ready Reaction Mix and a 15 µL reaction volume

Volume of BigDye Terminator v1.1 & v3.1 5X Buffer (µL) = 0.5 x [(15/2.5) - 2] = 2 µL

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

Can I dilute my BigDye Terminator reaction?

We provide the BigDye Terminator v1.1 & v3.1 5X Buffer to enable dilution of the BigDye Terminator Ready Reaction Mix. However, without optimization, diluting the BigDye Terminator Ready Reaction Mix may cause deterioration of sequencing quality. We cannot guarantee the performance of BigDye chemistry when it is diluted.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.