Product Image
Applied Biosystems™

3730/3730xl DNA Analyzer Sequencing Standards, BigDye™ Terminator v1.1

This kit contains four tubes of hsp69 standard sequence used for instrument and spectral calibrations. This material is used withRead more
Have Questions?
Catalog NumberQuantity
43367991 Kit
Catalog number 4336799
Price (USD)
2,065.00
Each
Quantity:
1 Kit
Price (USD)
2,065.00
Each
This kit contains four tubes of hsp69 standard sequence used for instrument and spectral calibrations. This material is used with the 3730, and 3730xl DNA Analyzers.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Sequencing
For Use With (Equipment)3730xl DNA Analyzer, 3730 DNA Analyzer
Label or DyeROX, TAMRA™, Rhodamine 110, Rhodamine 6G
Product LineBigDye Terminator, BigDye
Product TypeSequencing Standards
Quantity1 Kit
Shipping ConditionDry Ice
Unit SizeEach
Contents & Storage
This kit contains 4 tubes of hsp69 standard sequence used for instrument and spectral calibrations. This material is used with the 3730, and 3730xl DNA Analyzers. Store kit at -15 to -25°C.

Frequently asked questions (FAQs)

I see another sequence running under mine. What can cause this?

Some of the causes of another sequence appearing under the primary sequence are:

• Contamination: There is more than one species of DNA present (e.g., multiple PCR products).

• Primers: Primer is annealing in more than one location on the template, primer dimer, primer degradation or not manufactured properly resulting in N+1 or N-1, carry over from PCR reaction, or primers pairs in a multiplex reaction that are not appropriate for multiplexing (i.e., primers anneal inappropriately).

• Spectral/Matrix: If the raw data signal intensity of the sample is too high or saturated it can exceed the amount of color bleedthrough (or spectral overlap) that the matrix (310) or spectral (3130, 3730, 3500) are removing, resulting in secondary peaks appearing in a very specific pattern (e.g., a red peak always appearing under a green peak). A change in the camera, laser, or optical alignment requires that a new matrix be made or a new spectral calibration be performed.

For more information on more than one sequence or set of peaks in a sequencing run, please refer to the DNA Sequencing by Capillary Electrophoresis: Applied Biosystems Chemistry Guide: Second Edition (Cat. No. 4305080, Rev. C). The guide can be found by searching the Thermo Fisher Scientific website with the catalog number 4305080.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

How often should a spectral calibration be run on the Applied Biosystems 3730 Series DNA Analyzer?

There is no fixed time as to when a spectral must be run. However, some users prefer to run a new spectral at a preset time to ensure it is up to date. Data quality will be the largest indicator of when a new spectral is required. A separate spectral calibration needs to be performed for each dye set, array type, and array length. For G5-RCT dye set users only: a spectral calibration must be re-run if the array detection cell is moved/repositioned. Refer to FAQ "When should I run a spectral calibration?" for more information on when a spectral should be run.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.

When should I run a spectral calibration on the Applied Biosystems 3730 Series DNA Analyzer?

A new spectral calibration should be run:

- Whenever a new dye set is used on an instrument.
- After any optics adjustment performed by a service engineer.
- If the instrument has been moved.
- If you see a decrease in spectral separation (pull-up and/or pull-down peaks).
- If you alter any conditions (dye set, array type, or array length).
- For G5-RCT, Any4dye-HDR, and S dye set users only: Any time the array detection cell is moved/repositioned.



Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.

What is a spectral calibration?

A spectral calibration is an algorithm applied to raw data, which converts it into the component 4 or 5 dye data stored in the sample files. A spectral is created for a specific dye set (combination of dyes), array type (4 or 16 capillaries), and array length (36cm or 50cm). It is used to correct for the natural overlap of the fluorescent dyes.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.

What is the shelf life of the sequencing standard?

The sequencing standard has a shelf life of one year from the date of shipment, unless otherwise listed on the CofA. Our Terms and Conditions of sale can be found here(https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf).

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.