4-Methylbenzylmagnesium chloride, 0.5M solution in THF, AcroSeal™
4-Methylbenzylmagnesium chloride, 0.5M solution in THF, AcroSeal™
4-Methylbenzylmagnesium chloride, 0.5M solution in THF, AcroSeal™
Thermo Scientific Chemicals

4-Methylbenzylmagnesium chloride, 0.5M solution in THF, AcroSeal™

CAS: 29875-07-8 | C8H9ClMg | 164.92 g/mol
Have Questions?
Catalog NumberQuantityPackaging
43370050050 mLAcroSeal™ Glass Bottle
Catalog number 433700500
Price (USD)
-
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS29875-07-8
IUPAC Namechloro[(4-methylphenyl)methyl]magnesium
Molecular FormulaC8H9ClMg
InChI KeySAKYCLWKOWGTFT-UHFFFAOYSA-M
SMILESCC1=CC=C(C[Mg]Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to green to brown
Appearance (Form)Solution
Titration Grignard0.45 to 0.55 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only