Morpholine, 99.5%, purified by redistillation, AcroSeal™
Morpholine, 99.5%, purified by redistillation, AcroSeal™
Morpholine, 99.5%, purified by redistillation, AcroSeal™
Thermo Scientific Chemicals

Morpholine, 99.5%, purified by redistillation, AcroSeal™

CAS: 110-91-8 | C4H9NO | 87.12 g/mol
数量:
100 mL
800 mL
パッケージ:
AcroSeal™ Glass bottle
製品番号(カタログ番号) 433858000
価格(JPY)
-
数量:
800 mL
パッケージ:
AcroSeal™ Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS110-91-8
IUPAC Namemorpholine
Molecular FormulaC4H9NO
InChI KeyYNAVUWVOSKDBBP-UHFFFAOYSA-N
SMILESC1COCCN1
さらに表示
Appearance (Form)Clear liquid
Color scale=<20 APHA
Infrared spectrumConforms
Refractive index1.4530 to 1.4550 (20°C, 589 nm)
GC>=99.4 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only