GeneAmp™ PCR System 9700 and 9800 Fast Thermal Cycler, IQ User-Performed Test Procedures
Applied Biosystems™
GeneAmp™ PCR System 9700 and 9800 Fast Thermal Cycler, IQ User-Performed Test Procedures
In laboratories regulated typically by GxP and ISO, compliance with government and international standards requires documented verification that your systems are installed and functioning according to their operational specifications.
Have Questions?
Catalog NumberQuantity
43393861 each
Catalog number 4339386
Price (MXN)
-
Quantity:
1 each
In laboratories regulated typically by GxP and ISO, compliance with government and international standards requires documented verification that your systems are installed and functioning according to their operational specifications. This IQ (Installation Qualification) User-Performed Test Procedures package provides you with test procedures to document that your GeneAmp™ PCR System 9700 and 9800 Fast Thermal Cycler is installed to manufacturer's specifications.
The procedures should be performed when the instrument has been relocated or modifications have been made to the instrument's configuration.
This package includes a bound, printed copy of test procedures. Directions for performing the test procedures can be found in the instrument user manual. The package does not include Temperature Verification Kits and Probes.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineGeneAmp
Product TypeThermal Cycler IQ User-Performed Test Procedures
Quantity1 each
Unit SizeEach
Contents & Storage
Procedures and instructions for initial qualification of GeneAmp 9700 and 9800 PCR systems
Citations & References (2420)
Citations & References
Abstract
Analysis of bacterial DNA in synovial tissue of Tunisian patients with reactive and undifferentiated arthritis by broad-range PCR, cloning and sequencing
Authors:Siala, M; Jaulhac, B; Gdoura, R; Sibilia, J; Fourati, H; Younes, M; Baklouti, S; Bargaoui, N; Sellami, S; Znazen, A; Barthel, C; Collin, E; Hammami, A; Sghir, A
Journal:ARTHRITIS RESEARCH & THERAPY
PubMed ID:
Introduction Bacteria and/or their antigens have been implicated in the pathogenesis of reactive arthritis (ReA). Several studies have reported the presence of bacterial antigens and nucleic acids of bacteria other than those specified by diagnostic criteria for ReA in joint specimens from patients with ReA and various arthritides. The present ... More
Developmental Validation of Short Tandem Repeat Reagent Kit for Forensic DNA Profiling of Canine Biological Material
Authors:Dayton, M; Koskinen, MT; Tom, BK; Mattila, AM; Johnston, E; Halverson, J; Fantin, D; DeNise, S; Budowle, B; Smith, DG; Kanthaswamy, S
Journal:CROATIAN MEDICAL JOURNAL
PubMed ID:
Aim To develop a reagent kit that enables multiplex polymerase chain reaction (PCR) amplification of 18 short tandem repeats (STR) and the canine sex-determining Zinc Finger marker. Methods Validation studies to determine the robustness and reliability in forensic DNA typing of this multiplex assay included sensitivity testing, reproducibility studies, ... More
Phenotypic and genotypic diversity among strains of Aureobasidium pullulans in comparison with related species
Authors:Loncaric, I; Oberlerchner, JT; Heissenberger, B; Moosbeckhofer, R
Journal:Antonie Van Leeuwenhoek International Journal of General and Molecular Microbiology
PubMed ID:
Intra-specific diversity of 200 Aureobasidium pullulans strains isolated from different sources and their relatives Kabatiella lini CBS 125.21 T and Hormonema prunorum CBS 933.72 T were studied by assessment of macromorphological, and physiological tests, sodium dodecyl sulfate-polyacrylamide gel electrophoresis technique (SDS-PAGE) of whole-cell proteins as well as enterobacterial repetitive intergenic ... More
Identification of common traits in improved xylose-growing Saccharomyces cerevisiae for inverse metabolic engineering
Authors:Bengtsson, O; Jeppsson, M; Sonderegger, M; Parachin, NS; Sauer, U; Hahn-Haegerdal, B; Gorwa-Grauslund, MF
Journal:YEAST
PubMed ID:
Four recombinant Saccharomyces cerevisiae strains with enhanced xylose growth (TMB3400, C1, C5 and BH42) were compared with two control strains (TMB3399, TMB3001) through genome-wide transcription analysis in order to identify novel targets for inverse metabolic engineering. A subset of 13 genes with changed expression levels in all improved strains was ... More
Primary extragastrointestinal stromal tumor of the transverse mesocolon without c-kit mutations but with PDGFRA mutations
Authors:Terada, T
Journal:MEDICAL ONCOLOGY
PubMed ID:
The author reports a rare case of primary large cystic extragastrointestinal stromal tumor (eGIST) of the transverse mesocolon with genetic analyses of the c-kit and platelet-derived growth factor receptor-α (PDGFRA) genes. A 78-year-old man was found to have a large cystic tumor in the abdomen, and the tumor was resected. ... More