POP™ Conformational Analysis Polymer (CAP) - FAQs
POP™ Conformational Analysis Polymer (CAP) - FAQs
View additional product information for POP™ Conformational Analysis Polymer (CAP) - FAQs (4340379)
1 product FAQs found
Do you offer any fragment analysis-based kits for non-denaturing applications?
We provide POP Conformational Analysis Polymer (CAP) for non-denaturing polynucleotide fragment analysis applications on the Applied Biosystems 3130/3130xl Genetic Analyzers. However, we do not have a premade kit specifically for this application. To learn more about non-denaturing applications, please see here (http://www.thermofisher.com/us/en/home/life-science/sequencing/fragment-analysis/single-strand-conformation-polymorphism-sscp.html).
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.