Product Image
Applied Biosystems™

Acetic Anhydride/Pyridine/THF, Capping Formula A

This acetic anhydride/tetrahydrofolate/pyridine (Cap A) solution comes in Trap-Pak™ bags. Trap-Pak™ bags are tightly sealed membrane packets containing molecular sieves to efficiently adsorb water.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
43408654 x 180 mL
43408604 x 450 mL
43473814 x 2 L
Catalog number 4340865
Price (USD)
427.00
Each
Quantity:
4 x 180 mL
Price (USD)
427.00
Each

This acetic anhydride/tetrahydrofolate/pyridine (Cap A) solution comes in Trap-Pak™ bags. Trap-Pak™ bags are tightly sealed membrane packets containing molecular sieves to efficiently adsorb water. Use Trap-Pak™ bags to reduce and maintain a low water content in acetonitrile and tetrazole (activator) solutions.

  • Extensive testing and quality controls assure high quality reagents that meet our high expectations and specifications.
  • Reagents and supplies are use-tested and optimized for the 3900 system for quality nucleic acid synthesis.

Tested
Our high-performance reagents begin with raw materials that must conform to our strict specifications. Once the reagents are manufactured and packaged, extensive testing is performed to assure you that all liquids are free of interfering particulate matter; reagent concentrations are within our specifications, and the performance specification meets at least 98% stepwise coupling efficiency.

WARNING: Cancer – www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostics procedures.

Specifications
Chemical Name or MaterialAcetic anhydride/pyridine/tetrahydrofuran
For Use With (Application)Oligo Synthesis
For Use With (Equipment)3900 DNA Synthesizer
FormLiquid
Product TypeOligonucleotide Synthesis Reagents
Quantity4 x 180 mL
Reagent TypeSolvent or General Reagent
Sample TypeDNA
Unit SizeEach
Contents & Storage
4 x 180 mL